missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Aminobenzenesulphonic Acid, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Aniline-2-sulfonic acid |
| CAS | 88-21-1 |
| Molecular Formula | C6 H7 N O3 S |
| Quantity | 10 g |
| Synonym | Benzenesulfonic acid, o-amino- (6CI,7CI,8CI), 2-Aminobenzenesulfonic acid, 1-Aminobenzene-2-sulfonic acid, 2-Aminophenylsulfonic acid, 2-Anilinesulfonic acid, 2-Sulfoaniline, Aniline-o-sulfonic acid, NSC 147, Orthanilic acid, o-Aminobenzenesulfonic acid, o-Aminophenylsulfonic acid, o-Sulfanilic acid |
| InChI Formula | InChI=1S/C6H7NO3S/c7-5-3-1-2-4-6(5)11(8,9)10/h1-4H,7H2,(H,8,9,10) |
| SMILES | Nc1ccccc1S(=O)(=O)O |
| IUPAC Name | 2-aminobenzenesulfonic acid |
| Molecular Weight (g/mol) | 173.19 |
| Formula Weight | 173.0147 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction