missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzeneboronic acid, 96%

Product Code. 11394895
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394895 5 g 5g
11384895 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394895 Supplier Thermo Scientific Alfa Aesar Supplier No. L18069.06
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 5570-18-3 | C6H8BNO2 | 136.945 g/mol

2-Aminobenzeneboronic acid derivative is used in the synthesis of quinolines by reacting with alpha, beta unsaturated ketones. Its hydrochloride derivative is used in the preparation of designed boronate ligands for glucose-selective holographic sensors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzeneboronic acid derivative is used in the synthesis of quinolines by reacting with alpha, beta unsaturated ketones. Its hydrochloride derivative is used in the preparation of designed boronate ligands for glucose-selective holographic sensors.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5570-18-3
Molecular Formula C6H8BNO2
Molecular Weight (g/mol) 136.945
MDL Number MFCD01074645
InChI Key DIRRKLFMHQUJCM-UHFFFAOYSA-N
Synonym 2-aminophenyl boronic acid, 2-aminobenzeneboronic acid, o-aminophenylboronic acid, 2-aminophenylboronicacid, 2-boronoaniline, pubchem9885, acmc-1apjj, 2-amino-phenylboronic acid, 2-amino benzeneboronic acid
PubChem CID 2773216
IUPAC Name (2-aminophenyl)boronic acid
SMILES B(C1=CC=CC=C1N)(O)O

Specifications

Melting Point 188°C to 190°C
Quantity 5 g
Beilstein 2962939
Formula Weight 136.95
Percent Purity 96%
Chemical Name or Material 2-Aminobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.