missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzaldehyde, 97%

Product Code. 10351770 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10351770 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10351770 Supplier Thermo Scientific Acros Supplier No. 279660010
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 529-23-7 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 529-23-7
CAS Min % 96.0
CAS Max % 100.0
Melting Point 37°C
Color White to Yellow
Boiling Point 80°C to 85°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H7NO
Linear Formula NH2C6H4CHO
MDL Number MFCD00007709
Quantity 1 g
Synonym o-aminobenzaldehyde, benzaldehyde, 2-amino, 2-amino-benzaldehyde, aminobenzaldehyde, anthranilaldehyde, 2-formylaniline, anthranil aldehyde, o-amino benzaldehyde, benzaldehyde, amino, pubchem15692
InChI Key FXWFZIRWWNPPOV-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C=O)N
IUPAC Name 2-aminobenzaldehyde
Molecular Weight (g/mol) 121.14
PubChem CID 68255
Formula Weight 121.14
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 2-Aminobenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.