missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoanthracene, TRC

Product Code. 30318830
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318830 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318830 Supplier Toronto Research Chemicals Supplier No. TRCA576890500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Aminoanthracene
CAS 613-13-8
Molecular Formula C14 H11 N
Quantity 500 mg
Synonym 2-Anthracenamine (9CI, ACI), 2-Anthramine (6CI, 7CI, 8CI), (Anthracen-2-yl)amine, 2-Aminoanthracene, 2-Anthracylamine, 2-Anthrylamine, NSC 400535, β-Aminoanthracene
InChI Formula InChI=1S/C14H11N/c15-14-6-5-12-7-10-3-1-2-4-11(10)8-13(12)9-14/h1-9H,15H2
SMILES Nc1ccc2cc3ccccc3cc2c1
IUPAC Name anthracen-2-amine
Molecular Weight (g/mol) 193.24
Formula Weight 193.0891
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.