missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoadenosine, 97%

Product Code. 11429990
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429990 5 g 5g
11419990 1 g 1g
11439990 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429990 Supplier Thermo Scientific Alfa Aesar Supplier No. 044748.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2096-10-8 | C10H14N6O4 | 282.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2096-10-8
MDL Number MFCD00053556
ChEBI CHEBI:1014

Specifications

Molecular Formula C10H14N6O4
Synonym 2-aminoadenosine, 2-amine adenosine, adenosine, 2-amino, 2,6-diaminopurine riboside, 2,6-diaminonebularine, 2,6-diaminopurinosine, 2r,3r,4s,5r-2-2,6-diaminopurin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2,6-diamino-9-beta-d-ribofuranosyl purine, pubchem14156, ksc202m1p
InChI Key ZDTFMPXQUSBYRL-SBTUYZBDNA-N
SMILES NC1=NC(N)=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=N1
IUPAC Name (2R,3R,4S,5R)-2-(2,6-diaminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 282.26
Quantity 5 g
PubChem CID 72200
Formula Weight 282.26
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 2-Aminoadenosine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.