missing translation for 'onlineSavingsMsg'
Learn More

2-Aminoacetophenone hydrochloride, 97%

Product Code. 11402240
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11402240 1 g 1g
11412240 10 g 10g
11422240 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11402240 Supplier Thermo Scientific Alfa Aesar Supplier No. H29117.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5468-37-1 | C8H10ClNO | 171.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5468-37-1
Molecular Formula C8H10ClNO
Molecular Weight (g/mol) 171.62
MDL Number MFCD00012873
InChI Key CVXGFPPAIUELDV-UHFFFAOYSA-N
Synonym 2-aminoacetophenone hydrochloride, 2-amino-1-phenylethanone hydrochloride, phenacylamine hydrochloride, 2-aminoacetophenone hcl, 2-aminoacetophenonehydrochloride, 2-amino-1-phenylethan-1-one hydrochloride, alpha-aminoacetophenone hydrochloride, 2-aminoacetophenonehcl, ethanone, 2-amino-1-phenyl-, hydrochloride, omega-aminoacetophenone hydrochloride
PubChem CID 2723597
IUPAC Name 2-amino-1-phenylethanone;hydrochloride
SMILES Cl.NCC(=O)C1=CC=CC=C1

Specifications

Melting Point ∽194°C (decomposition)
Odor Odorless
Quantity 1 g
Formula Weight 171.63
Percent Purity 98%
Chemical Name or Material 2-Aminoacetophenone hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.