missing translation for 'onlineSavingsMsg'
Learn More

2'-Aminoacetophenone, 98%

Product Code. 11319216
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319216 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11319216 Supplier Thermo Scientific Alfa Aesar Supplier No. A10895.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 551-93-9 | C8H9NO | 135.166 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 551-93-9
Density 1.118
Boiling Point 253°C to 255°C
Flash Point 112°C
Molecular Formula C8H9NO
Refractive Index 1.616
MDL Number MFCD00007717
Quantity 10 g
Beilstein 386122
Merck Index 14,413
Synonym 2 '-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline
InChI Key GTDQGKWDWVUKTI-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=CC=C1N
IUPAC Name 1-(2-aminophenyl)ethanone
Molecular Weight (g/mol) 135.166
PubChem CID 11086
Formula Weight 135.17
Percent Purity 98%
Chemical Name or Material 2 '-Aminoacetophenone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.