missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride, 97%

Product Code. 15490636
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490636 1 g 1g
15400646 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15490636 Supplier Thermo Scientific Alfa Aesar Supplier No. H53398.03
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1171331-39-7 | C4H8ClF3N2O | 192.566 g/mol

Reactant in the synthesis of isoxazoline indolizine amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of isoxazoline indolizine amides.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1171331-39-7
Molecular Formula C4H8ClF3N2O
Molecular Weight (g/mol) 192.566
MDL Number MFCD09944516
InChI Key DBNFKWRZLGVLSH-UHFFFAOYSA-N
Synonym 2-amino-n-2,2,2-trifluoroethyl acetamide hydrochloride, 2-amino-n-2,2,2-trifluoroethyl acetamide;hydrochloride, c4h7f3n2o.hcl, 2-amino-n-2,2,2-trifluoroethyl acetamide hcl
PubChem CID 42913698
IUPAC Name 2-amino-N-(2,2,2-trifluoroethyl)acetamide;hydrochloride
SMILES C(C(=O)NCC(F)(F)F)N.Cl

Specifications

Linear Formula H2NCH2(CO)NHCH2CF3·HCl
Quantity 1 g
Formula Weight 192.57
Percent Purity 97%
Chemical Name or Material 2-Amino-N-(2,2,2-trifluoroethyl)acetamide hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.