missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-m-cresol, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-m-cresol |
| CAS | 2835-97-4 |
| Molecular Formula | C7H9NO |
| Quantity | 2.5 g |
| Synonym | 2-Amino-3-methylphenol, 2-Amino-3-hydroxytoluene, 2-Hydroxy-3-methylaniline, 2-Hydroxy-6-methylaniline, 3-Methyl-2-aminophenol |
| InChI Formula | InChI=1S/C7H9NO/c1-5-3-2-4-6(9)7(5)8/h2-4,9H,8H2,1H3 |
| SMILES | OC1=CC=CC(C)=C1N |
| IUPAC Name | 2-Amino-3-methylphenol; 2-Amino-3-hydroxytoluene; 2-Hydroxy-3-methylaniline; 2-Hydroxy-6-methylaniline; 3-Methyl-2-aminophenol |
| Molecular Weight (g/mol) | 123.15 |
| Formula Weight | 123.07 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction