missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-7-bromofluorene, tech. 90%

Product Code. 15471588
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471588 5 g 5g
15461588 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471588 Supplier Thermo Scientific Alfa Aesar Supplier No. H55370.06
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6638-60-4 | C13H10BrN | 260.134 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Sparingly soluble in water. (0.724 mg/L)

Notes
Incompatible with oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6638-60-4
Molecular Formula C13H10BrN
Molecular Weight (g/mol) 260.134
MDL Number MFCD00001127
InChI Key RJWYTISHBMNMOZ-UHFFFAOYSA-N
Synonym 2-amino-7-bromofluorene, 2-bromo-7-aminofluorene, fluoren-2-amine, 7-bromo, 7-bromo-9h-fluoren-2-ylamine, 9h-fluoren-2-amine, 7-bromo, 7-bromofluorene-2-ylamine, acmc-20a5iw, 2-amino-7-bromo-9h-fluorene, 9h-fluoren-2-amine,7-bromo, 7-bromanyl-9h-fluoren-2-amine
PubChem CID 23126
IUPAC Name 7-bromo-9H-fluoren-2-amine
SMILES C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br

Specifications

Melting Point 148°C to 149°C
Quantity 5 g
Solubility Information Sparingly soluble in water. (0.724mg/L)
Formula Weight 260.13
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Amino-7-bromofluorene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.