Learn More
2-Amino-6-(trifluoromethoxy)benzothiazole, 98%, Thermo Scientific Chemicals
£67.60 - £180.00
Chemical Identifiers
CAS | 1744-22-5 |
---|---|
Molecular Formula | C8H5F3N2OS |
Molecular Weight (g/mol) | 234.196 |
MDL Number | MFCD00210213 |
InChI Key | FTALBRSUTCGOEG-UHFFFAOYSA-N |
Synonym | riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanish |
PubChem CID | 5070 |
ChEBI | CHEBI:8863 |
IUPAC Name | 6-(trifluoromethoxy)-1,3-benzothiazol-2-amine |
SMILES | C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15457858
|
Thermo Scientific Alfa Aesar
H61328.06 |
5 g |
£67.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15467858
|
Thermo Scientific Alfa Aesar
H61328.14 |
25 g |
£180.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1744-22-5 | |
234.196 | |
FTALBRSUTCGOEG-UHFFFAOYSA-N | |
5070 | |
6-(trifluoromethoxy)-1,3-benzothiazol-2-amine |
C8H5F3N2OS | |
MFCD00210213 | |
riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanish | |
CHEBI:8863 | |
C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N |
Specifications
1744-22-5 | |
White to Yellow | |
MFCD00210213 | |
748759 | |
FTALBRSUTCGOEG-UHFFFAOYSA-N | |
6-(trifluoromethoxy)-1,3-benzothiazol-2-amine | |
5070 | |
234.2 | |
Solid |
114°C to 118°C | |
C8H5F3N2OS | |
UN2811 | |
riluzole, rilutek, 2-amino-6-trifluoromethoxy benzothiazole, 6-trifluoromethoxy benzo d thiazol-2-amine, 6-trifluoromethoxy-1,3-benzothiazol-2-amine, riluzolum, riluzol, 2-amino-6-trifluoromethoxybenzothiazole, riluzole hcl, riluzol inn-spanish | |
C1=CC2=C(C=C1OC(F)(F)F)SC(=N2)N | |
234.196 | |
CHEBI:8863 | |
98% | |
2-Amino-6-(trifluoromethoxy)benzothiazole |
Safety and Handling
GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P310-P330-P501c
H300
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packaging Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'rtecsNumber' : DL2830000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only