missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-6-purinethiol, 95%, ACROS Organics™
£25.70 - £80.10
Chemical Identifiers
CAS | 154-42-7 |
---|---|
Molecular Formula | C5H5N5S |
Molecular Weight (g/mol) | 167.19 |
MDL Number | MFCD00233553 |
InChI Key | WYWHKKSPHMUBEB-UHFFFAOYSA-N |
Synonym | 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol |
PubChem CID | 2723601 |
ChEBI | CHEBI:9555 |
IUPAC Name | 2-amino-6,7-dihydro-3H-purine-6-thione |
SMILES | NC1=NC(=S)C2=C(N1)N=CN2 |
Description
Chemical Identifiers
154-42-7 | |
167.19 | |
WYWHKKSPHMUBEB-UHFFFAOYSA-N | |
2723601 | |
2-amino-6,7-dihydro-3H-purine-6-thione |
C5H5N5S | |
MFCD00233553 | |
6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol | |
CHEBI:9555 | |
NC1=NC(=S)C2=C(N1)N=CN2 |
Specifications
154-42-7 | |
100.0 | |
Yellow to Green | |
95% | |
MFCD00233553 | |
15, 9490 | |
Solubility in water: soluble in water. Other solubilities: soluble in an aqueous naoh solution | |
NC1=NC(=S)C2=C(N1)N=CN2 | |
167.19 | |
CHEBI:9555 | |
95% | |
2-Amino-6-purinethiol |
94.0 | |
>360.0°C | |
Authentic | |
C5H5N5S | |
26, IV, 3926 | |
6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol | |
WYWHKKSPHMUBEB-UHFFFAOYSA-N | |
2-amino-6,7-dihydro-3H-purine-6-thione | |
2723601 | |
167.19 | |
Powder |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Amino-6-purinethiol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Germ cell mutagenicity Category 2
Hazard Statement
- H341-Suspected of causing genetic defects.
- H301-Toxic if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /