missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-purinethiol, 95%, ACROS Organics™

Product Code. 10419780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10419780 1 g Glass bottle 1g
10771345 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10419780 Supplier Thermo Scientific Acros Supplier No. 104480010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 154-42-7
Molecular Formula C5H5N5S
Molecular Weight (g/mol) 167.19
MDL Number MFCD00233553
InChI Key WYWHKKSPHMUBEB-UHFFFAOYSA-N
Synonym 6-thioguanine, thioguanine, tioguanine, 2-amino-6-mercaptopurine, 6-mercaptoguanine, tioguanin, tabloid, lanvis, 2-amino-6-purinethiol, 2-aminopurine-6-thiol
PubChem CID 2723601
ChEBI CHEBI:9555
IUPAC Name 2-amino-6,7-dihydro-3H-purine-6-thione
SMILES NC1=NC(=S)C2=C(N1)N=CN2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >360.0°C
Color Yellow to Green
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 1 g
Beilstein 26, IV, 3926
Merck Index 15, 9490
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in an aqueous naoh solution
Formula Weight 167.19
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 2-Amino-6-purinethiol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-6-purinethiol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.