missing translation for 'onlineSavingsMsg'
Learn More

(-)-2-Amino-6-propionamido-tetrahydrobenzothiazole, TRC

Product Code. 30012654
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012654 100 mg 100mg
30009736 50 mg 50mg
30008103 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012654 Supplier Toronto Research Chemicals Supplier No. TRCA628150100MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106006-84-2
Molecular Formula C10 H15 N3 O S
Molecular Weight (g/mol) 225.31
Synonym N-[(6S)-2-Amino-4,5,6,7-tetrahydro-6-benzothiazolyl]propanamide, (S)-N-(2-Amino-4,5,6,7-tetrahydro-6-benzothiazolyl)propanamide
IUPAC Name N-[(6S)-2-amino-4,5,6,7-tetrahydro-1,3-benzothiazol-6-yl]propanamide
SMILES CCC(=O)N[C@H]1CCc2nc(N)sc2C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H15N3OS/c1-2-9(14)12-6-3-4-7-8(5-6)15-10(11)13-7/h6H,2-5H2,1H3,(H2,11,13)(H,12,14)/t6-/m0/s1
Formula Weight 225.0936
Percent Purity >95%
Chemical Name or Material (-)-2-Amino-6-propionamido-tetrahydro-benzothiazole
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.