missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methylbenzothiazole, 99%

Product Code. 11435397
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11435397 100 g 100g
11425397 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435397 Supplier Thermo Scientific Alfa Aesar Supplier No. B23422.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2536-91-6 | C8H8N2S | 164.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2536-91-6
Molecular Formula C8H8N2S
Molecular Weight (g/mol) 164.23
MDL Number MFCD00005789
InChI Key DZWTXWPRWRLHIL-UHFFFAOYSA-N
Synonym 2-amino-6-methylbenzothiazole, 6-methylbenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methyl, 6-methyl-2-aminobenzothiazole, 2-methylbenzothiazol-2-amine, 6-methyl-2-benzothiazolylamine, benzothiazole, 2-amino-6-methyl, 6-methyl-benzothiazol-2-ylamine, 2-amino-6-methyl-benzothiazole, 6-methyl-2-benzothiazolamine
PubChem CID 17335
IUPAC Name 6-methyl-1,3-benzothiazol-2-amine
SMILES CC1=CC=C2N=C(N)SC2=C1

Specifications

Melting Point 139°C to 142°C
Quantity 100 g
Formula Weight 164.22
Percent Purity 99%
Physical Form Crystals or Powder or Crystalline Powder
Chemical Name or Material 2-Amino-6-methylbenzothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.