missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methyl-4(3H)-quinazolone, 96%, Thermo Scientific Chemicals

Product Code. 11333586
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11333586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11333586 Supplier Thermo Scientific Alfa Aesar Supplier No. L14558.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50440-82-9
Molecular Formula C9H9N3O
Molecular Weight (g/mol) 175.191
MDL Number MFCD00236015
InChI Key DJYDUJAKTAWKTN-UHFFFAOYSA-N
Synonym 2-amino-6-methylquinazolin-4 3h-one, 2-amino-6-methyl-4 3h-quinazolone, 2-amino-6-methyl-3h-quinazolin-4-one, 2-amino-6-methyl-4 3h-quinazolinone, acmc-1aogs, chembl7222, 2-amino-4-hydroxy-6-methylquinazoline, 2-amino-6-methylquinazolin-4 1h-one, 6-methyl-2-aminoquinazolin-4 3h-one, 2-amino-6-methyl-3-hydroquinazolin-4-one
PubChem CID 4151444
IUPAC Name 2-amino-6-methyl-1H-quinazolin-4-one
SMILES CC1=CC2=C(C=C1)NC(=NC2=O)N

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 609421
Solubility Information Slightly soluble in water.
Formula Weight 175.19
Percent Purity 96%
Chemical Name or Material 2-Amino-6-methyl-4(3H)-quinazolone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.