missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-methoxybenzothiazole, 98%

Product Code. 10256130 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10256130 50 g Glass bottle 50g
10201932 5 g Glass bottle 5g
10791251 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10256130 Supplier Thermo Scientific Acros Supplier No. 172020500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1747-60-0 | C8H8N2OS | 180.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1747-60-0
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.23
MDL Number MFCD00005787
InChI Key KZHGPDSVHSDCMX-UHFFFAOYSA-N
Synonym 2-amino-6-methoxybenzothiazole, 6-methoxybenzo d thiazol-2-amine, 2-benzothiazolamine, 6-methoxy, 6-methoxy-2-aminobenzothiazole, 2-amino-6-methoxy benzothiazole, benzothiazole, 2-amino-6-methoxy, 6-methoxy-2-benzothiazolamine, 6-methoxybenzothiazol-2-ylamine, unii-lat2842yvx, ccris 1393
PubChem CID 15630
IUPAC Name 6-methoxy-1,3-benzothiazol-2-amine
SMILES COC1=CC2=C(C=C1)N=C(S2)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 162°C to 169°C
Color Beige to White or Gray-Lilac
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 50 g
Beilstein 27, II, 334
Formula Weight 180.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-methoxybenzothiazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-6-methoxybenzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.