missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-fluorobenzoic acid, 98%

Product Code. 11404313
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404313 5 g 5g
11494303 1 g 1g
11414313 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11404313 Supplier Thermo Scientific Alfa Aesar Supplier No. A10919.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 434-76-4 | C7H5FNO2 | 154.12 g/mol

Used in the synthesis of a novel benzamide with potent neuroleptic activity. 5- fluoro-2-methyl-3,l-benzoxazin-4-one was obtained from 2-amino-6-fluorobenzoic acid. The condensation of 2-amino-6-fluorobenzoic acid methyl ester and (4-fluorophenyl) acetyl chloride with N,N-dimethylamino-4-pyridine in CH 2 Cl 2 to give 2-fluoro-6-[2-(4-fluoro-phenyl)-acetylamino]-benzoic acid methyl ester in 61% yield. 2 Amino 6 fluorobenzoic acid is another easily ac cessible synthon for construction of monofluorinated heterocycles. Reactions of 2,3,4,6-tetra-O-acetyl-n-D_glucopyranosyl bromide (2) with p-hydroxybenzaldehyde, trifluoromethyl and fluoro substi- tuted aniline and 2-amino-6-fluorobenzoic acid gave lO-(Pformyl- pheny1)-2,3,4,6-tetra-O-acetyl-(3-D-glucopyranoside. 2-Fluoro-6-iodobenzoic acid,a loose white crystal,was synthesized by using 2-amino-6-fluorobenzoic acid as the starting material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of a novel benzamide with potent neuroleptic activity. 5- fluoro-2-methyl-3,l-benzoxazin-4-one was obtained from 2-amino-6-fluorobenzoic acid. The condensation of 2-amino-6-fluorobenzoic acid methyl ester and (4-fluorophenyl) acetyl chloride with N,N-dimethylamino-4-pyridine in CH 2 Cl 2 to give 2-fluoro-6-[2-(4-fluoro-phenyl)-acetylamino]-benzoic acid methyl ester in 61% yield. 2 Amino 6 fluorobenzoic acid is another easily ac cessible synthon for construction of monofluorinated heterocycles. Reactions of 2,3,4,6-tetra-O-acetyl-n-D_glucopyranosyl bromide (2) with p-hydroxybenzaldehyde, trifluoromethyl and fluoro substi- tuted aniline and 2-amino-6-fluorobenzoic acid gave lO-(Pformyl- pheny1)-2,3,4,6-tetra-O-acetyl-(3-D-glucopyranoside. 2-Fluoro-6-iodobenzoic acid,a loose white crystal,was synthesized by using 2-amino-6-fluorobenzoic acid as the starting material.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 434-76-4
Molecular Formula C7H5FNO2
Molecular Weight (g/mol) 154.12
MDL Number MFCD00067781
InChI Key RWSFZKWMVWPDGZ-UHFFFAOYSA-M
Synonym 6-fluoroanthranilic acid, anthralic acid, 6-fluoro, benzoic acid, 2-amino-6-fluoro, 6-amino-2-fluorobenzoic acid, 2-amino-6-fluoro-benzoic acid, 2,amino-6-fluorobenzoic acid, 4owo, pubchem1324, 6-fluoroanthanilic acid, acmc-209jun
PubChem CID 521142
IUPAC Name 2-amino-6-fluorobenzoic acid
SMILES NC1=CC=CC(F)=C1C([O-])=O

Specifications

Melting Point 165°C to 167°C
Quantity 5 g
Beilstein 3540926
Formula Weight 155.13
Percent Purity 98%
Chemical Name or Material 2-Amino-6-fluorobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.