missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-ethylpyridine, 97%

Product Code. 11419436
Change view
Click to view available options
Quantity:
2.5 g
10 g
50 g
Unit Size:
10g
2.5g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419436 2.5 g 2.5g
11429436 10 g 10g
11439436 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419436 Supplier Thermo Scientific Alfa Aesar Supplier No. A19146.88
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 21717-29-3 | C7H10N2 | 122.171 g/mol

The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The action of iodine on 2-amino-6-ethylpyridine in an alkaline medium gives 2-amino-5-iodo-6-ethylpyridine.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21717-29-3
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
MDL Number MFCD00218265
InChI Key JXKAUUVMXZIJNZ-UHFFFAOYSA-N
Synonym 2-amino-6-ethylpyridine, 6-ethyl-2-pyridinamine, 2-pyridinamine, 6-ethyl, 6-ethyl-2-pyridylamine, acmc-1clku, 2-pyridinamine,6-ethyl, 2-amino-6-ethyl-pyridine, 6-ethyl-pyridin-2-ylamine
PubChem CID 89021
IUPAC Name 6-ethylpyridin-2-amine
SMILES CCC1=NC(=CC=C1)N

Specifications

Odor Unpleasant
Quantity 2.5 g
UN Number UN2810
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material 2-Amino-6-ethylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.