missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropyridine, 97%, Thermo Scientific Chemicals

Catalog Number 10649579
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
45644-21-1
C5H5ClN2
128.56
MFCD00234068
OBYJTLDIQBWBHM-UHFFFAOYSA-N
2-amino-6-chloropyridine, 2-chloro-6-aminopyridine, 2-pyridinamine, 6-chloro, 6-chloro-2-pyridinamine, 6-chloro-pyridin-2-ylamine, pyridine, 2-amino-6-chloro, 6-chloropyridin-2-ylamine, 6-chloro-2-pyridylamine, 6-amino-2-chloropyridine, 2-amino-6-chloropyridine 6-chloropyridin-2-ylamine
206246
6-chloropyridin-2-amine
C1=CC(=NC(=C1)Cl)N
This item is not returnable. View return policy
45644-21-1
C5H5ClN2
128.56
MFCD00234068
OBYJTLDIQBWBHM-UHFFFAOYSA-N
2-amino-6-chloropyridine, 2-chloro-6-aminopyridine, 2-pyridinamine, 6-chloro, 6-chloro-2-pyridinamine, 6-chloro-pyridin-2-ylamine, pyridine, 2-amino-6-chloro, 6-chloropyridin-2-ylamine, 6-chloro-2-pyridylamine, 6-amino-2-chloropyridine, 2-amino-6-chloropyridine 6-chloropyridin-2-ylamine
206246
6-chloropyridin-2-amine
C1=CC(=NC(=C1)Cl)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 72.0°C to 73.5°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 128.56
Percent Purity 97%
Physical Form Powder or Low Melting Solid
Chemical Name or Material 2-Amino-6-chloropyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-6-chloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'