missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloropurine, 99+%, Thermo Scientific Chemicals

Product Code. 10183190 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183190 5 g Glass bottle 5g
10624622 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10183190 Supplier Thermo Scientific Acros Supplier No. 149290050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10310-21-1
Molecular Formula C5H4ClN5
Molecular Weight (g/mol) 169.57
MDL Number MFCD00075252
InChI Key RYYIULNRIVUMTQ-UHFFFAOYSA-N
Synonym 2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine
PubChem CID 5360349
ChEBI CHEBI:72345
IUPAC Name 6-chloro-7H-purin-2-amine
SMILES NC1=NC(Cl)=C2NC=NC2=N1

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point >300°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: 1.7g/L water (20°C)
Formula Weight 169.57
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 2-Amino-6-chloropurine, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-6-chloropurine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.