Learn More
2-Amino-6-chloropurine, 99%, Thermo Scientific Chemicals
£36.70 - £228.00
Chemical Identifiers
CAS | 10310-21-1 |
---|---|
Molecular Formula | C5H4ClN5 |
Molecular Weight (g/mol) | 169.57 |
MDL Number | MFCD00075252 |
InChI Key | RYYIULNRIVUMTQ-UHFFFAOYSA-N |
Synonym | 2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine |
PubChem CID | 5360349 |
ChEBI | CHEBI:72345 |
IUPAC Name | 6-chloro-7H-purin-2-amine |
SMILES | NC1=NC(Cl)=C2NC=NC2=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11483717
|
Thermo Scientific Alfa Aesar
A18195.03 |
1 g |
£36.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11493717
|
Thermo Scientific Alfa Aesar
A18195.06 |
5 g |
£77.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11403727
|
Thermo Scientific Alfa Aesar
A18195.14 |
25 g |
£228.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2?-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-6-chloropurine finds its uses in the enzymatic synthesis of 2′-deoxyguanosine, 9-alkyl purines and (R)- and (S)-N-(2-phosphonomethoxypropyl) derivatives of purine and pyrimidine bases.
Solubility
Solubility in 1mol/L NaOH; very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Identificadores de productos químicos
10310-21-1 | |
169.57 | |
RYYIULNRIVUMTQ-UHFFFAOYSA-N | |
5360349 | |
6-chloro-7H-purin-2-amine |
C5H4ClN5 | |
MFCD00075252 | |
2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine | |
CHEBI:72345 | |
NC1=NC(Cl)=C2NC=NC2=N1 |
Especificaciones
10310-21-1 | |
C5H4ClN5 | |
9626 | |
RYYIULNRIVUMTQ-UHFFFAOYSA-N | |
6-chloro-7H-purin-2-amine | |
5360349 | |
169.57 | |
2-Amino-6-chloropurine |
>300°C | |
MFCD00075252 | |
2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine | |
NC1=NC(Cl)=C2NC=NC2=N1 | |
169.57 | |
CHEBI:72345 | |
99% |
Seguridad y manipulación
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H360
missing translation for 'einecsNumber' : 233-686-7
missing translation for 'rtecsNumber' : UO7502000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only