missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzothiazole, 99%, Thermo Scientific™

Product Code. 10477150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10477150 10 g Glass bottle 10g
10361134 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10477150 Supplier Thermo Scientific Acros Supplier No. 136400100
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 95-24-9
Molecular Formula C7H5ClN2S
Molecular Weight (g/mol) 184.65
MDL Number MFCD00053557
InChI Key VMNXKIDUTPOHPO-UHFFFAOYSA-N
Synonym 2-amino-6-chlorobenzothiazole, 6-chlorobenzothiazol-2-ylamine, 2-benzothiazolamine, 6-chloro, 6-chlorobenzo d thiazol-2-amine, 6-chloro-benzothiazol-2-ylamine, unii-2u337t5ufg, 6-chloro-2-benzothiazolamine, benzothiazole, 2-amino-6-chloro, 6-chlorobenzothiazole-2-ylamine, c7h4brcln2s
PubChem CID 7226
IUPAC Name 6-chloro-1,3-benzothiazol-2-amine
SMILES C1=CC2=C(C=C1Cl)SC(=N2)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 197°C to 201°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 10 g
Formula Weight 184.65
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-6-chlorobenzothiazole
Show More Show Less
compliance-icons
Product Identifier
  • 2-Amino-6-chlorobenzothiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.