missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chlorobenzoic acid, 97+%, Thermo Scientific Chemicals

Product Code. 11474243
Click to view available options
Quantity:
5 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-6-chlorobenzoic acid is used as intermediate for medicine.

Solubility
Very soluble in water. Soluble in methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2148-56-3
Molecular Formula C7H6ClNO2
Molecular Weight (g/mol) 171.58
MDL Number MFCD00051530
InChI Key SZCPTRGBOVXVCA-UHFFFAOYSA-N
Synonym 6-chloroanthranilic acid, benzoic acid, 2-amino-6-chloro, 2-amino-6-chloro-benzoic acid, 6-chloranthranilsaure, pubchem9876, acmc-1cngq, 6-chloro anthranilic acid, 2-carboxy-3-chloroaniline, 2amino-6-chlorobenzoic acid, ksc201s6b
PubChem CID 75071
IUPAC Name 2-amino-6-chlorobenzoic acid
SMILES C1=CC(=C(C(=C1)Cl)C(=O)O)N

Specifications

Melting Point 149°C to 152°C
Quantity 25 g
Beilstein 2804041
Solubility Information Very soluble in water. Soluble in methanol.
Formula Weight 171.58
Percent Purity 98%
Chemical Name or Material 2-Amino-6-chlorobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'