missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-chloro-4-(diethylamino)pyrimidine, Thermo Scientific™

Product Code. 10331710
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10331710 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10331710 Supplier Thermo Scientific Maybridge Supplier No. MAY00271DA
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 116895-09-1
Packaging Amber Glass Bottle
Molecular Formula C8H13ClN4
MDL Number MFCD00664696
Quantity 1 g
Synonym 2-amino-6-chloro-4-diethylamino pyrimidine, 6-chloro-n4,n4-diethylpyrimidine-2,4-diamine, 6-chloro-n∼4∼,n∼4∼-diethyl-2,4-pyrimidinediamine, 6-chloro-n*4*,n*4*-diethyl-pyrimidine-2,4-diamine, 2-amino-6-chloropyrimidin-4-yl diethylamine, acmc-20alz2, 6-chloro-n∼4∼,n∼4∼-diethylpyrimidine-2,4-diamine, 2,4-pyrimidinediamine, 6-chloro-n∼4∼,n∼4∼-diethyl, 2,4-pyrimidinediamine,6-chloro-n4,n4-diethyl-diethyl, 6-chloro-n4,n4-diethyl-pyrimidine-2,4-diamine
InChI Key JZPXSIGRVGTURL-UHFFFAOYSA-N
SMILES CCN(CC)C1=CC(Cl)=NC(N)=N1
IUPAC Name 6-chloro-4-N,4-N-diethylpyrimidine-2,4-diamine
Molecular Weight (g/mol) 200.67
PubChem CID 1484287
Formula Weight 200.67
Percent Purity 97%
Chemical Name or Material 2-amino-6-chloro-4-(diethylamino)pyrimidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.