missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromopyridine, 98%

Product Code. 15720869 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15720869 5 g Glass bottle 5g
10082282 25 g Glass bottle 25g
11926331 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15720869 Supplier Thermo Scientific Acros Supplier No. 376870050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19798-81-3 | C5H6BrN2 | 174.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19798-81-3
Molecular Formula C5H6BrN2
Molecular Weight (g/mol) 174.02
MDL Number MFCD00137843
InChI Key BKLJUYPLUWUEOQ-UHFFFAOYSA-O
Synonym 2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663
PubChem CID 300809
IUPAC Name 6-bromopyridin-2-amine
SMILES NC1=CC=CC(Br)=[NH+]1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88.0°C to 91.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 173.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-6-bromopyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.