missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromopyridine, 98%

Product Code. 11485980
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11485980 25 g 25g
11475980 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485980 Supplier Thermo Scientific Alfa Aesar Supplier No. H26555.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 19798-81-3 | C5H6BrN2 | 174.02 g/mol

Employed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed in an efficient one-pot synthesis of 7-azaindoles. Used in the synthesis of anti-HIV agents.

Solubility
Slightly soluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19798-81-3
Molecular Formula C5H6BrN2
Molecular Weight (g/mol) 174.02
MDL Number MFCD00137843
InChI Key BKLJUYPLUWUEOQ-UHFFFAOYSA-O
Synonym 2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663
PubChem CID 300809
IUPAC Name 6-bromopyridin-2-amine
SMILES NC1=CC=CC(Br)=[NH+]1

Specifications

Melting Point 88°C to 91°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 173.01
Percent Purity 98%
Chemical Name or Material 2-Amino-6-bromopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.