missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6-bromopyridine, 98%

Product Code. 10082282 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10082282 25 g Glass bottle 25g
15720869 5 g Glass bottle 5g
11926331 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10082282 Supplier Thermo Scientific Acros Supplier No. 376870250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19798-81-3 | C5H6BrN2 | 174.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19798-81-3
Molecular Formula C5H6BrN2
Molecular Weight (g/mol) 174.02
MDL Number MFCD00137843
InChI Key BKLJUYPLUWUEOQ-UHFFFAOYSA-O
Synonym 2-amino-6-bromopyridine, 6-bromo-2-pyridinamine, aminobromopyridine, 6-bromo-2-aminopyridine, 6-bromo-pyridin-2-ylamine, 2-pyridinamine, 6-bromo, 6-bromo-2-pyridylamine, 2-amino-6-bromo-pyridine, zlchem 805, pubchem7663
PubChem CID 300809
IUPAC Name 6-bromopyridin-2-amine
SMILES NC1=CC=CC(Br)=[NH+]1

Specifica

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88.0°C to 91.0°C
Color Brown to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 173.01
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-6-bromopyridine
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 2-Amino-6-bromopyridine
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.