missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-6,8-dihydroxypurine Hydrochloride (~90%), TRC

Product Code. 30012452
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012452 100 mg 100mg
30011312 10 mg 10mg
30003234 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012452 Supplier Toronto Research Chemicals Supplier No. TRCA604921100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246818-54-1
Molecular Formula C5H6ClN5O2
Molecular Weight (g/mol) 203.59
Synonym 2-Amino-7,9-dihydro-1H-purine-6,8-dione Hydrochloride, 2-Amino-6,8-purinediol Hydrochloride, 8-Hydroxyguanine Hydrochloride, 8-Oxo-7,8-dihydroguanine Hydrochloride, 8-Oxoguanine Hydrochloride, NSC 22720;
IUPAC Name 2-amino-8-hydroxy-1,7-dihydropurin-6-one;hydrochloride
SMILES C12=C(NC(=N)N=C1O)NC(=N2)O.Cl

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H5N5O2.ClH/c6-4-8-2-1(3(11)10-4)7-5(12)9-2;/h(H5,6,7,8,9,10,11,12);1H
Formula Weight 203.021
Chemical Name or Material 2-Amino-6,8-dihydroxypurine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.