missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrothiazole, TRC

Product Code. 30137780
Change view
Click to view available options
Quantity:
1 g
50 g
100 g
Unit Size:
100g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137780 50 g 50g
30134281 100 g 100g
17923705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137780 Supplier Toronto Research Chemicals Supplier No. TRCA61865050G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-66-4
Molecular Formula C3 H3 N3 O2 S
Molecular Weight (g/mol) 145.14
Synonym 2-Thiazolamine, 5-nitro-, Thiazole, 2-amino-5-nitro- (6CI,7CI,8CI), 2-Amino-4-nitrothiazole, 2-Amino-5-nitro-1,3-thiazole, 2-Amino-5-nitrothiazole, 5-Nitro-2-aminothiazole, 5-Nitro-2-thiazolamine, 5-Nitro-2-thiazolylamine, Amnizol soluble, Enheptin, Enheptin T, Entramin, NSC 4, Nitramin IDO
IUPAC Name 5-nitro-1,3-thiazol-2-amine
SMILES Nc1ncc(s1)[N+](=O)[O-]

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H3N3O2S/c4-3-5-1-2(9-3)6(7)8/h1H,(H2,4,5)
Formula Weight 144.9946
Chemical Name or Material 2-Amino-5-nitrothiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.