missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrophenol, 95%

Product Code. 10229740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
250g
25g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10229740 250 g Glass bottle 250g
10552294 25 g Glass bottle 25g
10340664 50 g Glass bottle 50g
10455085 1 kg Glass bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10229740 Supplier Thermo Scientific Acros Supplier No. 400902500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 121-88-0 | C6H6N2O3 | 154.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-88-0
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.13
MDL Number MFCD00007692
InChI Key DOPJTDJKZNWLRB-UHFFFAOYSA-N
Synonym 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene
PubChem CID 4984721
ChEBI CHEBI:82384
IUPAC Name 2-amino-5-nitrophenol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)N

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 198.0°C to 202.0°C
Color Brown to Brown-Rust
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 250 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, benzene and diethylether
Formula Weight 154.13
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 2-Amino-5-nitrophenol, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Amino-5-nitrophenol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.