missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzotrifluoride, 98%, Thermo Scientific™

Product Code. 10154060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10154060 25 g 25g
10072440 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10154060 Supplier Thermo Scientific Acros Supplier No. 188230250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 121-01-7
MDL Number MFCD00007365
InChI Key HOTZLWVITTVZGY-UHFFFAOYSA-N
Synonym 2-amino-5-nitrobenzotrifluoride, 4-nitro-2-trifluoromethyl aniline, 2-amino-5-nitro-trifluorobenzene, 4-nitro-alpha,alpha,alpha-trifluoro-o-toluidine, 4-nitro-2-trifluoromethyl benzenamine, 4-nitro-a,a,a-trifluoro-o-toluidine, benzenamine, 4-nitro-2-trifluoromethyl, 5-nitro-2-aminobenzotrifluoride, 2-trifluoromethyl-4-nitroaniline, a,a,a-trifluoro-4-nitro-o-toluidine
PubChem CID 67128
IUPAC Name 4-nitro-2-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1[N+](=O)[O-])C(F)(F)F)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 90°C to 93°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H5F3N2O2
Linear Formula H2NC6H3(NO2)CF3
Quantity 25 g
Molecular Weight (g/mol) 206.124
Formula Weight 206.12
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material 2-Amino-5-nitrobenzotrifluoride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.