missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7080717
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
616-79-5
C7H6N2O4
182.14
MFCD00017039
RUCHWTKMOWXHLU-UHFFFAOYSA-N
5-nitroanthranilic acid, benzoic acid, 2-amino-5-nitro, 2-amino-5-nitro-benzoic acid, 2-amino-5-nitrobenzoicacid, anthranilic acid, 5-nitro, 1-amino-2-carboxy-4-nitrobenzene, 2-amino-5-nitro-benzoicacid, pubchem14422, acmc-209mvd, 5-nitro anthranilic acid
12032
CHEBI:61280
2-amino-5-nitrobenzoic acid
NC1=CC=C(C=C1C(O)=O)[N+]([O-])=O
This item is not returnable. View return policy
616-79-5
C7H6N2O4
182.14
MFCD00017039
RUCHWTKMOWXHLU-UHFFFAOYSA-N
5-nitroanthranilic acid, benzoic acid, 2-amino-5-nitro, 2-amino-5-nitro-benzoic acid, 2-amino-5-nitrobenzoicacid, anthranilic acid, 5-nitro, 1-amino-2-carboxy-4-nitrobenzene, 2-amino-5-nitro-benzoicacid, pubchem14422, acmc-209mvd, 5-nitro anthranilic acid
12032
CHEBI:61280
2-amino-5-nitrobenzoic acid
NC1=CC=C(C=C1C(O)=O)[N+]([O-])=O

2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY
Melting Point ∼270°C (decomposition)
Quantity 10 g
Beilstein 646219
Solubility Information Insoluble in water.
Formula Weight 182.14
Percent Purity 98%
Chemical Name or Material 2-Amino-5-nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'