missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitrobenzoic acid, 98%

Product Code. 11438633
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438633 250 g 250g
11418633 10 g 10g
11428633 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438633 Supplier Thermo Scientific Alfa Aesar Supplier No. A10426.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 616-79-5 | C7H6N2O4 | 182.14 g/mol

2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-nitrobenzoic acid, is used as an intermediate for dyes and pharmaceuticals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to strong oxidizing agents. Stable under ordinary conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-79-5
Molecular Formula C7H6N2O4
Molecular Weight (g/mol) 182.14
MDL Number MFCD00017039
InChI Key RUCHWTKMOWXHLU-UHFFFAOYSA-N
Synonym 5-nitroanthranilic acid, benzoic acid, 2-amino-5-nitro, 2-amino-5-nitro-benzoic acid, 2-amino-5-nitrobenzoicacid, anthranilic acid, 5-nitro, 1-amino-2-carboxy-4-nitrobenzene, 2-amino-5-nitro-benzoicacid, pubchem14422, acmc-209mvd, 5-nitro anthranilic acid
PubChem CID 12032
ChEBI CHEBI:61280
IUPAC Name 2-amino-5-nitrobenzoic acid
SMILES NC1=CC=C(C=C1C(O)=O)[N+]([O-])=O

Specifications

Melting Point ∼270°C (decomposition)
Quantity 250 g
Beilstein 646219
Solubility Information Insoluble in water.
Formula Weight 182.14
Percent Purity 98%
Chemical Name or Material 2-Amino-5-nitrobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.