missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-nitro-4-picoline, 98%, Thermo Scientific™

Product Code. 10003650 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10003650 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10003650 Supplier Thermo Scientific Acros Supplier No. 319560010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 21901-40-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 220.0°C to 225.0°C
Color Brown-Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H7N3O2
MDL Number MFCD00010692
Quantity 1 g
Synonym 2-amino-4-methyl-5-nitropyridine, 2-amino-5-nitro-4-picoline, 2-amino-4-methyl-5-nitro pyridine, 4-methyl-5-nitro-2-pyridinamine, 4-methyl-5-nitro-2-pyridylamine, 2-pyridinamine, 4-methyl-5-nitro, 4-methyl-5-nitro-pyridin-2-ylamine, 2-amino-5-nitro-4-methylpyridine, 4-methyl-5-nitropyridin-2-amin, 2-amino-5-nitro-4-picoline 2-amino-4-methyl-5-nitropyridine
InChI Key GRBBNZYMXKTQAI-UHFFFAOYSA-N
SMILES CC1=CC(=NC=C1[N+](=O)[O-])N
IUPAC Name 4-methyl-5-nitropyridin-2-amine
Molecular Weight (g/mol) 153.14
PubChem CID 243165
Formula Weight 153.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-5-nitro-4-picoline, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.