missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylthiazole, 98%

Product Code. 10095913 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10095913 100 g Glass bottle 100g
10530554 5 g Glass bottle 5g
10106753 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10095913 Supplier Thermo Scientific Acros Supplier No. 392791000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 7305-71-7 | C4H6N2S | 114.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7305-71-7
Molecular Formula C4H6N2S
Molecular Weight (g/mol) 114.17
MDL Number MFCD00078317
InChI Key GUABFMPMKJGSBQ-UHFFFAOYSA-N
Synonym 2-amino-5-methylthiazole, 5-methylthiazol-2-amine, 2-thiazolamine, 5-methyl, 2-amino-5-methyl-thiazole, thiazole, 2-amino-5-methyl, 5-methyl-2-thiazolamine, 5-methyl-2-aminothiazole, unii-7rab78af2p, 2-amino-5-methyl thiazole, meloxicam related compound b
PubChem CID 351770
IUPAC Name 5-methyl-1,3-thiazol-2-amine
SMILES CC1=CN=C(S1)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 93.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 114.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Amino-5-methylthiazole
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Amino-5-methylthiazole
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.