missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methylpyridine, 99%

Product Code. 11470943
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470943 10 g 10g
11480943 50 g 50g
11490943 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470943 Supplier Thermo Scientific Alfa Aesar Supplier No. A13330.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1603-41-4 | C6H8N2 | 108.144 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1603-41-4
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006328
InChI Key CMBSSVKZOPZBKW-UHFFFAOYSA-N
Synonym 2-amino-5-methylpyridine, 2-amino-5-picoline, 6-amino-3-picoline, 2-pyridinamine, 5-methyl, 5-methylpyridin-2-ylamine, 3-picoline, 6-amino, 5-methyl-2-pyridinamine, 5-methyl-2-pyridylamine, 5-methyl-2-aminopyridine, 2-amino-5-methyl pyridine
PubChem CID 15348
IUPAC Name 5-methylpyridin-2-amine
SMILES CC1=CN=C(C=C1)N

Specifications

Melting Point 74°C to 78°C
Boiling Point 226°C to 228°C
Flash Point 118°C (244°F)
Odor Unpleasant
Quantity 10 g
UN Number UN2811
Beilstein 107050
Formula Weight 108.14
Percent Purity 99%
Chemical Name or Material 2-Amino-5-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.