missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-Methylhexane Hydrochloride, TRC

Product Code. 30132474
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132474 10 g 10g
30005052 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132474 Supplier Toronto Research Chemicals Supplier No. TRCA61622210G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71776-71-1
Molecular Formula C7H17N . HCl
Molecular Weight (g/mol) 151.68
Synonym (±)-2-Amino-5-methylhexane Hydrochloride, 1,4-Dimethylpentylamine Hydrochloride, 5-Methyl-2-hexylamine Hydrochloride, dl-2-Amino-5-methylhexane Hydrochloride;
IUPAC Name 5-methylhexan-2-amine hydrogen Chloride
SMILES CC(CCC(N)C)C.Cl

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H17N.ClH/c1-6(2)4-5-7(3)8;/h6-7H,4-5,8H2,1-3H3;1H
Formula Weight 151.12
Chemical Name or Material 2-Amino-5-Methylhexane Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.