missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-methyl-1,3,4-thiadiazole, 97%

Product Code. 10307240 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10307240 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10307240 Supplier Thermo Scientific Acros Supplier No. 158370050
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 108-33-8 | C3H5N3S | 115.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 108-33-8
CAS Min % 96.0
CAS Max % 100.0
Melting Point 224°C to 225°C
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C3H5N3S
MDL Number MFCD00003110
Quantity 5 g
Beilstein 27, 629
Synonym 2-amino-5-methyl-1,3,4-thiadiazole, 2-amino-5-methylthiadiazole, matd, usaf cy-3, 1,3,4-thiadiazol-2-amine, 5-methyl, 5-methyl-1,3,4 thiadiazol-2-ylamine, 1,3,4-thiadiazole, 2-amino-5-methyl, 5-methyl-1,3,4-thiadiazole-2-amine, 5-methyl-1,3,4-thiadiazol-2-ylamine, 1,4-thiadiazol-2-amine, 5-methyl
InChI Key HMPUHXCGUHDVBI-UHFFFAOYSA-N
SMILES CC1=NN=C(N)S1
IUPAC Name 5-methyl-1,3,4-thiadiazol-2-amine
Molecular Weight (g/mol) 115.15
PubChem CID 66949
Formula Weight 115.16
Percent Purity 97%
Chemical Name or Material 2-Amino-5-methyl-1, 3, 4-thiadiazole, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.