missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodobenzoic Acid, TRC

Product Code. 30340061
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340061 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340061 Supplier Toronto Research Chemicals Supplier No. TRCA6305131G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-iodobenzoic Acid
CAS 5326-47-6
Molecular Formula C7H6INO2
Quantity 1 g
Synonym Anthranilic acid, 5-iodo- (6CI,8CI), 5-Iodo-2-aminobenzoic acid, 5-Iodoanthranilic acid, NSC 302
InChI Formula InChI=1S/C7H6INO2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3H,9H2,(H,10,11)
SMILES Nc1ccc(I)cc1C(=O)O
IUPAC Name 2-amino-5-iodobenzoic acid
Molecular Weight (g/mol) 263.03
Formula Weight 262.9443
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.