missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-iodo-3-methylbenzoic Acid Methyl Ester, TRC

Product Code. 30327176
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327176 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327176 Supplier Toronto Research Chemicals Supplier No. TRCA60411525G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-iodo-3-methylbenzoic Acid Methyl Ester
CAS 1096481-48-9
Molecular Formula C9 H10 I N O2
Quantity 25 g
InChI Formula InChI=1S/C9H10INO2/c1-5-3-6(10)4-7(8(5)11)9(12)13-2/h3-4H,11H2,1-2H3
SMILES COC(=O)c1cc(I)cc(C)c1N
IUPAC Name methyl 2-amino-5-iodo-3-methylbenzoate
Molecular Weight (g/mol) 291.086
Formula Weight 290.976
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.