missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-5-formylthiazole, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2-Amino-5-formylthiazole |
| CAS | 1003-61-8 |
| Molecular Formula | C4 H4 N2 O S |
| Quantity | 10 mg |
| Synonym | 2-Amino-5-thiazolecarboxaldehyde, 2-Amino-1,3-thiazole-5-carboxaldehyde |
| InChI Formula | InChI=1S/C4H4N2OS/c5-4-6-1-3(2-7)8-4/h1-2H,(H2,5,6) |
| SMILES | N=C1NC=C(S1)C=O |
| IUPAC Name | 2-imino-3H-1,3-thiazole-5-carbaldehyde |
| Molecular Weight (g/mol) | 128.152 |
| Formula Weight | 128.004 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction