missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluoropyridine, 97%

Product Code. 11394396
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
missing translation for 'unitSize'
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394396 5 g 5g
11384396 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 11394396 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' L20010.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21717-96-4 | C5H5FN2 | 112.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21717-96-4
Molecular Formula C5H5FN2
Molecular Weight (g/mol) 112.11
MDL Number MFCD01861120
InChI Key YJTXQLYMECWULH-UHFFFAOYSA-N
Synonym 2-amino-5-fluoropyridine, 5-fluoro-2-pyridinamine, 5-fluoro-2-pyridineamine, 5-fluoro-pyridin-2-ylamine, 5-fluoro-2-pyridylamine, 2-amino 5-fluoropyridine, 2-pyridinamine, 5-fluoro, 5-fluoro-2-aminopyridine, 2-amino-5-fluoro pyridine, 5-fluoropyridimine
PubChem CID 2737701
IUPAC Name 5-fluoropyridin-2-amine
SMILES NC1=CC=C(F)C=N1

Specifications

Melting Point 93°C to 97°C
Odor Characteristic
Quantity 5 g
UN Number UN2811
Beilstein 385938
Formula Weight 112.11
Percent Purity 97%
Chemical Name or Material 2-Amino-5-fluoropyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.