missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluorobenzoic acid, 97%

Product Code. p-7025786
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11426974

Brand: Thermo Scientific Alfa Aesar B21458.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 446-08-2 | C7H6FNO2 | 155.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-08-2
Molecular Formula C7H6FNO2
Molecular Weight (g/mol) 155.128
MDL Number MFCD00055566
InChI Key FPQMGQZTBWIHDN-UHFFFAOYSA-N
Synonym 5-fluoroanthranilic acid, 2-amino-5-fluoro-benzoic acid, benzoic acid, 2-amino-5-fluoro, 5-faa, 5-fluoro-2-aminobenzoic acid, 4own, 5rg, pubchem1322, 5-fluoroanthanilic acid, acmc-209jyo
PubChem CID 101412
ChEBI CHEBI:78042
IUPAC Name 2-amino-5-fluorobenzoic acid
SMILES C1=CC(=C(C=C1F)C(=O)O)N

Specifications

Melting Point 182°C to 184°C
Quantity 1 g
Beilstein 2803664
Formula Weight 155.13
Percent Purity ≥98%
Chemical Name or Material 2-Amino-5-fluorobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.