missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluoro-6-methylpyridine, 98%

Product Code. 15462009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462009 5 g 5g
15452009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462009 Supplier Thermo Scientific Alfa Aesar Supplier No. H64700.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 110919-71-6 | C6H7FN2 | 126.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110919-71-6
Molecular Formula C6H7FN2
Molecular Weight (g/mol) 126.13
MDL Number MFCD09835298
InChI Key WMGRXNKQEVOVET-UHFFFAOYSA-N
Synonym 2-amino-5-fluoro-6-methylpyridine, 5-fluoro-6-methyl-2-pyridinamine, 2-pyridinamine, 5-fluoro-6-methyl, 2-amino-5-fluoro-6-methyl-pyridine, 5-fluoro-6-methyl-pyridin-2-ylamine, 5-fluoro-6-methyl-2-pyridylamine, 6-amino-3-fluoro-2-methylpyridine
PubChem CID 13789841
IUPAC Name 5-fluoro-6-methylpyridin-2-amine
SMILES CC1=C(F)C=CC(N)=N1

Specifications

Color White
Quantity 5 g
UN Number UN2811
Beilstein 4376910
Formula Weight 126.13
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Amino-5-fluoro-6-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.