missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-fluoro-3-nitropyridine, 98%, Thermo Scientific Chemicals

Product Code. 15477808
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 212268-12-7
Molecular Formula C5H4FN3O2
Molecular Weight (g/mol) 157.10
MDL Number MFCD04972413
InChI Key LDYYBZNEWDTDEE-UHFFFAOYSA-N
Synonym 2-amino-3-nitro-5-fluoropyridine, 2-amino-5-fluoro-3-nitropyridine, 5-fluoro-3-nitro-2-pyridinamine, pubchem1271, acmc-1cegr, ksc496c2h, 5-fluoro-3-nitro-2-aminopyridine, 5-fluoro-3-nitropyridine-2-amine, 5-fluoro-3-nitro-pyridin-2-amine, 5-fluoro-3-nitro-2-pyridylamine
PubChem CID 21717339
IUPAC Name 5-fluoro-3-nitropyridin-2-amine
SMILES NC1=NC=C(F)C=C1[N+]([O-])=O

Specifications

Color Orange
Quantity 1 g
Formula Weight 157.1
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 2-Amino-5-fluoro-3-nitropyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'