missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyclopropyl-1,3,4-thiadiazole, 98%

Product Code. 11317595
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11317595 1 g 1g
11327595 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11317595 Supplier Thermo Scientific Alfa Aesar Supplier No. L05778.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57235-50-4 | C5H7N3S | 141.19 g/mol

2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-cyclopropyl-1,3,4-thiadiazole is used as antibacterial agent.

Solubility
Soluble in Methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container. Store at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57235-50-4
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.19
MDL Number MFCD00051649
InChI Key AVLUMBXGKFNNAS-UHFFFAOYSA-N
Synonym 2-amino-5-cyclopropyl-1,3,4-thiadiazole, 5-cyclopropyl 1,3,4 thiadiazol-2-ylamine, 5-cyclopropyl-1,3,4-thiadiazole-2-ylamine, acmc-20ao4n, enamine_005503, 5-cyclopropyl-2-amino-1,3,4-thiadiazole, 1,3,4-thiadiazol-2-amine,5-cyclopropyl, 5-cyclopropyl-1,3,4 thiadiazol-2-ylamine
PubChem CID 717021
IUPAC Name 5-cyclopropyl-1,3,4-thiadiazol-2-amine
SMILES NC1=NN=C(S1)C1CC1

Specifications

Melting Point 212°C to 216°C
Quantity 1 g
Beilstein 124356
Solubility Information Soluble in Methanol. Insoluble in water.
Formula Weight 141.2
Percent Purity 98%
Chemical Name or Material 2-Amino-5-cyclopropyl-1,3,4-thiadiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.