missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-cyanopyridine, 98%

Product Code. 11354166
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354166 25 g 25g
11344166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354166 Supplier Thermo Scientific Alfa Aesar Supplier No. L19282.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4214-73-7 | C6H6N3 | 120.13 g/mol

2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Amino-5-cyanopyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4214-73-7
Molecular Formula C6H6N3
Molecular Weight (g/mol) 120.13
MDL Number MFCD00128902
InChI Key KDVBYUUGYXUXNL-UHFFFAOYSA-O
Synonym 2-amino-5-cyanopyridine, 6-aminonicotinonitrile, 6-amino-3-pyridinecarbonitrile, 6-amino-nicotinonitrile, 3-pyridinecarbonitrile, 6-amino, 2-amino-5-cyano pyridine, 6-aminonicotinonitril, zlchem 518, pubchem5752, acmc-209jnu
PubChem CID 818260
SMILES NC1=CC=C(C=[NH+]1)C#N

Specifications

Melting Point 159°C to 164°C
Boiling Point 240°C to 250°C (15 mmHg)
Quantity 25 g
UN Number UN3439
Beilstein 113876
Solubility Information Soluble in methanol. Insoluble in water.
IUPAC Name 6-aminopyridine-3-carbonitrile
Formula Weight 119.13
Percent Purity 98%
Chemical Name or Material 2-Amino-5-cyanopyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.