missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chloropyridine, Mikromol™

Product Code. 17972306
Change view
Click to view available options
Quantity:
25 mg
250 mg
Unit Size:
250mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972306 250 mg 250mg
17973456 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17972306 Supplier Mikromol Supplier No. MM0149.04
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Amino-5-chloropyridine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Amino-5-chloropyridine
CAS 1072-98-6
Analyte or Component Names 2-Amino-5-chloropyridine
Shipping Condition Room Temperature
Molecular Formula C5 H5 Cl N2
Certifications/Compliance ISO 17025
Synonym 2-Amino-5-chloropyridine
InChI Formula InChI=1S/C5H5ClN2/c6-4-1-2-5(7)8-3-4/h1-3H,(H2,7,8)
SMILES Nc1ccc(Cl)cn1
IUPAC Name 5-chloropyridin-2-amine
Molecular Weight (g/mol) 128.56
Quantity 250 mg
Formula Weight 128.0141 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.