missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-chlorobenzyl alcohol, 98%

Product Code. 15493947
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493947 25 g 25g
15483947 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493947 Supplier Thermo Scientific Alfa Aesar Supplier No. H30531.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 37585-25-4 | C7H8ClNO | 157.597 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37585-25-4
Molecular Formula C7H8ClNO
Molecular Weight (g/mol) 157.597
MDL Number MFCD00075163
InChI Key CLKBZWDZDVOIGJ-UHFFFAOYSA-N
PubChem CID 586754
IUPAC Name (2-amino-5-chlorophenyl)methanol
SMILES C1=CC(=C(C=C1Cl)CO)N

Specifications

Melting Point 107°C to 109°C
Quantity 25 g
Sensitivity Air Sensitive
Synonym 2-amino-5-chlorophenyl methanol, 2-amino-5-chlorobenzyl alcohol, 2-amino-5-chloro-phenyl-methanol, benzenemethanol, 2-amino-5-chloro, acmc-20ao4o, 2-amino-5-chlorobenzenemethanol, 2-hydroxymethyl-4-chloroaniline, benzenemethanol,2-amino-5-chloro, 2-amino-5-chlorophenyl methanol #
Formula Weight 157.6
Percent Purity 98%
Chemical Name or Material 2-Amino-5-chlorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.